Learn More
Medchemexpress LLC PF-4191834 50mg | 1029317-21-2 | 393.50 g·mol⁻¹ | C22H23N3O2S | 50 MG
SDP
Catalog No. 5000308329
Encompass_Preferred
PF-4191834 is an orally active, non-iron-chelating, non-redox inhibitor of 5-lipoxygenase (5-LOX) used in research to probe leukotriene biosynthesis and 5-LOX-mediated pathways. It is supplied as a solid, characterized by high purity, and has documented solubility and storage recommendations for both powder and solution forms.
  • Submicromolar potency against 5-LOX (reported IC50 ≈ 229 nM).
  • High purity (reported 99.77%).
  • Soluble in DMSO at approximately 100 mg/mL; in vivo formulation protocols documented.
  • Stable as powder when stored at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small research pack sizes including 50 mg.
Catalog No. 50-003-08329 Supplier Medchemexpress LLC Supplier No. HY11704850MG
PF-4191834 is an orally active, non-iron-chelating, non-redox inhibitor of 5-lipoxygenase (5-LOX) used in research to probe leukotriene biosynthesis and 5-LOX-mediated pathways. It is supplied as a solid, characterized by high purity, and has documented solubility and storage recommendations for both powder and solution forms.
  • Submicromolar potency against 5-LOX (reported IC50 ≈ 229 nM).
  • High purity (reported 99.77%).
  • Soluble in DMSO at approximately 100 mg/mL; in vivo formulation protocols documented.
  • Stable as powder when stored at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small research pack sizes including 50 mg.