Learn More
Medchemexpress LLC HY-18312 5mg , PF-4618433 CAS:1166393-85-6 Purity:>98%
SDP
Catalog No. 502260216
Encompass_Preferred
HY-18312 5mg PF-4618433 CAS: 1166393-85-6 PF-4618433 is a potent and selective PYK2 inhibitor, with an IC50 of 637 nM. PF-4618433 may be suitable for the research of osteoporosis, craniofacial and appendicular skeletal defects and for targeted bone regeneration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0216 Supplier Medchemexpress LLC Supplier No. HY183125MG
HY-18312 5mg PF-4618433 CAS: 1166393-85-6 PF-4618433 is a potent and selective PYK2 inhibitor, with an IC50 of 637 nM. PF-4618433 may be suitable for the research of osteoporosis, craniofacial and appendicular skeletal defects and for targeted bone regeneration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.