Learn More
Medchemexpress LLC HY-15773 100mg , PF-4708671 CAS:1255517-76-0 Purity:>98%
SDP
Catalog No. 502259259
Encompass_Preferred
HY-15773 100mg PF-4708671 CAS: 1255517-76-0 PF-4708671 is a potent cell-permeable S6K1 inhibitor with a Ki of 20 nM and IC50 of 160 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9259 Supplier Medchemexpress LLC Supplier No. HY15773100MG
HY-15773 100mg PF-4708671 CAS: 1255517-76-0 PF-4708671 is a potent cell-permeable S6K1 inhibitor with a Ki of 20 nM and IC50 of 160 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.