Learn More
Apexbio Technology LLC PF-4708671 1255517-76-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061801
Encompass_Preferred
PF-4708671 (CAS 1255517-76-0) is a cell-permeable small molecule that specifically inhibits p70 ribosomal S6 kinase 1 (S6K1) S6K1 regulates diverse cellular processes including protein synthesis and insulin signaling pathways PF-4708671 inhibits S6K1 with an IC50 of 160 nM in HEK293 cell-derived enzyme assays and has a reported Ki value of 20 nM in vitro It selectively inhibits S6K1 over structurally related kinases such as RSK2 and MSK1 hindering phosphorylation of downstream substrates including ribosomal protein S6 mTOR and Rictor PF-4708671 is valuable in biomedical research involving cancer and insulin resistance
Catalog No. 50-000-61801 Supplier Apexbio Technology LLC Supplier No. B22285.1
PF-4708671 (CAS 1255517-76-0) is a cell-permeable small molecule that specifically inhibits p70 ribosomal S6 kinase 1 (S6K1) S6K1 regulates diverse cellular processes including protein synthesis and insulin signaling pathways PF-4708671 inhibits S6K1 with an IC50 of 160 nM in HEK293 cell-derived enzyme assays and has a reported Ki value of 20 nM in vitro It selectively inhibits S6K1 over structurally related kinases such as RSK2 and MSK1 hindering phosphorylation of downstream substrates including ribosomal protein S6 mTOR and Rictor PF-4708671 is valuable in biomedical research involving cancer and insulin resistance