Learn More
Medchemexpress LLC HY-100176 1mg Medchemexpress, PF-4878691 CAS:532959-63-0 Purity:>98%
SDP
Catalog No. 501967714
Encompass_Preferred
Medchemexpress, HY-100176 1mg PF-4878691 CAS:532959-63-0 PF-4878691 (3M-852A) is a potent, orally active, and selective Toll-like receptor 7 (TLR7) agonist modelled to dissociate its antiviral and inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7714 Supplier Medchemexpress LLC Supplier No. HY1001761MG
Medchemexpress, HY-100176 1mg PF-4878691 CAS:532959-63-0 PF-4878691 (3M-852A) is a potent, orally active, and selective Toll-like receptor 7 (TLR7) agonist modelled to dissociate its antiviral and inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.