Learn More
Medchemexpress LLC HY-13864 5mg Medchemexpress, PF-4989216 CAS:1276553-09-3 Purity:>98%
SDP
Catalog No. 502081526
Encompass_Preferred
Medchemexpress, HY-13864 5mg PF-4989216 CAS:1276553-09-3 PF-4989216 is a potent and selective PI3Kα inhibitor with a Ki of 0.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1526 Supplier Medchemexpress LLC Supplier No. HY138645MG
Medchemexpress, HY-13864 5mg PF-4989216 CAS:1276553-09-3 PF-4989216 is a potent and selective PI3Kα inhibitor with a Ki of 0.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.