Learn More
Medchemexpress LLC HY-12584 5mg Medchemexpress, PF-5190457 CAS:1334782-79-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY125845MG
Encompass_Preferred
Medchemexpress, HY-12584 5mg PF-5190457 CAS:1334782-79-4 PF-5190457 is a potent and selective ghrelin receptor inverse agonist with a pKi of 8.36. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9702