Learn More
Medchemexpress LLC HY-13459 5mg Medchemexpress, PF-5274857 CAS:1373615-35-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134595MG
Encompass_Preferred
Medchemexpress, HY-13459 5mg PF-5274857 CAS:1373615-35-0 PF-5274857 is a potent, selective, orally active and brain-penetrant antagonist of Smo, with an IC50 of 5.8 nM and Ki of 4.6 nM. PF-5274857 has potential for research of tumor types including brain tumors and brain metastasis driven by an activated Hh pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1370