Learn More
Apexbio Technology LLC PF-543, 5mg. Cas: 1415562-82-1 MFCD: MFCD23098794. SphK1 inhibitor,cell-permeate,potent and selective
SDP
Supplier:  Apexbio Technology LLC A37175
Encompass_Preferred
PF-543 is a novel and cell-permeant inhibitor of Sphingosine kinase (SphK1) with IC50 value of 3.6 nM [1],SphK1 is a kinase that phosphorylates sphingosine to sphingosine-1-phosphate (SIP). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4830