Learn More
TARGETMOL CHEMICALS INC PF-543 HYDROCHLORIDE 10MG
SDP
Catalog No. 502576223
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-543 hydrochloride (PF-543) inhibits SphK1 with a K(i) of 3.6 nM is sphingosine-competitive and is more than 100-fold selective for SphK1 over the SphK2 isoform. purity: 99%
Catalog No. 50-257-6223 Supplier TARGETMOL CHEMICALS INC Supplier No. T884010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-543 hydrochloride (PF-543) inhibits SphK1 with a K(i) of 3.6 nM is sphingosine-competitive and is more than 100-fold selective for SphK1 over the SphK2 isoform. purity: 99%