Learn More
Selleck Chemical LLC PF-543 hydrochloride
SDP
Catalog No. 5000004516
Encompass_Preferred
PF-543 hydrochloride a novel sphingosine-competitive inhibitor of SphK1 inhibits SphK1 with IC50 and Ki of 2 0 nM and 3 6 nM exhibits 100-fold selectivity over the SphK2 isoform PF-543 hydrochloride induces apoptosis necrosis and autophagy
Catalog No. 50-000-04516 Supplier Selleck Chemical LLC Supplier No. S71771G
PF-543 hydrochloride a novel sphingosine-competitive inhibitor of SphK1 inhibits SphK1 with IC50 and Ki of 2 0 nM and 3 6 nM exhibits 100-fold selectivity over the SphK2 isoform PF-543 hydrochloride induces apoptosis necrosis and autophagy