Learn More
Medchemexpress LLC HY-10459 10mg , PF-562271 CAS:717907-75-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1045910MG
Encompass_Preferred
HY-10459 10mg PF-562271 CAS: 717907-75-0 PF-562271 (VS-6062) is a potent, ATP-competitive and reversible FAK and Pyk2 kinase inhibitor with IC50s of 1.5 nM and 13 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9618