Learn More
TARGETMOL CHEMICALS INC PF-562271 5MG
SDP
Catalog No. 502577115
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-562271 is an effective ATP-competitive reversible inhibitor of FAK(IC50=1.5 nM) and Pyk2 kinase(IC50=13 nM). purity: 97%
Catalog No. 50-257-7115 Supplier TARGETMOL CHEMICALS INC Supplier No. T24655MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-562271 is an effective ATP-competitive reversible inhibitor of FAK(IC50=1.5 nM) and Pyk2 kinase(IC50=13 nM). purity: 97%