Learn More
Selleck Chemical LLC PF-562271 Besylate S2672-1g
SDP
Catalog No. 503112866
Encompass_Preferred
PF-562271 Besylate is the benzenesulfonate salt of PF-562271 which is a potent ATP-competitive reversible inhibitor of FAK with IC50 of 1 5 nM 10-fold less potent for Pyk2 than FAK and 100-fold selectivity against other protein kinases except for some CDKs Phase 1
Catalog No. 50-311-2866 Supplier Selleck Chemical LLC Supplier No. S26721G
PF-562271 Besylate is the benzenesulfonate salt of PF-562271 which is a potent ATP-competitive reversible inhibitor of FAK with IC50 of 1 5 nM 10-fold less potent for Pyk2 than FAK and 100-fold selectivity against other protein kinases except for some CDKs Phase 1