Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-562271 besylate | 939791-38-5 | 99.2% | 665.66 g·mol⁻¹ | C27H26F3N7O6S2 | 100 MG
SDP

Catalog No. 5000243405
Encompass_Preferred

PF-562271 besylate is an ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 that exhibits low-nanomolar potency and is used in biochemical and cellular research to probe FAK/Pyk2 signaling pathways.

  • Potent inhibition of FAK and Pyk2 with IC50s of 1.5 nM and 13 nM.
  • High purity suitable for research applications.
  • Chemical formula C27H26F3N7O6S2; molecular weight 665.66 g·mol⁻¹.
  • Available in multiple package sizes, including 100 mg.
  • Storage: sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-43405 Supplier Medchemexpress LLC Supplier No. HY10458100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-562271 besylate is an ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 that exhibits low-nanomolar potency and is used in biochemical and cellular research to probe FAK/Pyk2 signaling pathways.

  • Potent inhibition of FAK and Pyk2 with IC50s of 1.5 nM and 13 nM.
  • High purity suitable for research applications.
  • Chemical formula C27H26F3N7O6S2; molecular weight 665.66 g·mol⁻¹.
  • Available in multiple package sizes, including 100 mg.
  • Storage: sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.