Learn More
Medchemexpress LLC PF-562271 besylate | 939791-38-5 | MFCD14105612 | 99.2% | 665.66 | C27H26F3N7O6S2 | 10 MG
SDP
Catalog No. 5000244036
Encompass_Preferred
PF-562271 besylate is the besylate salt of PF-562271, a potent ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 used in cancer research to probe signaling pathways, tumor growth, and cell adhesion. The compound is supplied as a high-purity solid for in vitro and in vivo studies.
  • Potent inhibition of FAK and Pyk2 (FAK IC50 ~1.5 nM; Pyk2 IC50 ~13 nM).
  • High reported purity (99.17%).
  • Provided as a solid, off-white to light-yellow material suitable for handling and formulation.
  • Demonstrated antiproliferative activity in multiple human cell lines.
  • Used for in vitro and in vivo studies of tumor growth, signaling, and angiogenesis.
  • Available in a 10 mg package for research use.
Catalog No. 50-002-44036 Supplier Medchemexpress LLC Supplier No. HY1045810MG
PF-562271 besylate is the besylate salt of PF-562271, a potent ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 used in cancer research to probe signaling pathways, tumor growth, and cell adhesion. The compound is supplied as a high-purity solid for in vitro and in vivo studies.
  • Potent inhibition of FAK and Pyk2 (FAK IC50 ~1.5 nM; Pyk2 IC50 ~13 nM).
  • High reported purity (99.17%).
  • Provided as a solid, off-white to light-yellow material suitable for handling and formulation.
  • Demonstrated antiproliferative activity in multiple human cell lines.
  • Used for in vitro and in vivo studies of tumor growth, signaling, and angiogenesis.
  • Available in a 10 mg package for research use.