Learn More
Medchemexpress LLC PF-562271 besylate | 939791-38-5 | MFCD14105612 | 99.2% | 665.66 | C27H26F3N7O6S2 | 50 MG
SDP
Catalog No. 5000244434
Encompass_Preferred
PF-562271 besylate is the besylate salt of a potent, ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 commonly used in preclinical biochemical and cellular research. It shows low-nanomolar activity against FAK and is supplied at high purity in small-scale packages for laboratory studies.
  • Potent ATP-competitive inhibitor of FAK and Pyk2 (FAK IC50 ≈ 1.5 nM; Pyk2 IC50 ≈ 13 nM).
  • Reversible mechanism suitable for biochemical assays.
  • High purity for research applications (reported ~99.17%).
  • Available in multiple small-scale package sizes for laboratory use.
  • Suitable for cellular and signaling pathway studies.
Catalog No. 50-002-44434 Supplier Medchemexpress LLC Supplier No. HY1045850MG
PF-562271 besylate is the besylate salt of a potent, ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 commonly used in preclinical biochemical and cellular research. It shows low-nanomolar activity against FAK and is supplied at high purity in small-scale packages for laboratory studies.
  • Potent ATP-competitive inhibitor of FAK and Pyk2 (FAK IC50 ≈ 1.5 nM; Pyk2 IC50 ≈ 13 nM).
  • Reversible mechanism suitable for biochemical assays.
  • High purity for research applications (reported ~99.17%).
  • Available in multiple small-scale package sizes for laboratory use.
  • Suitable for cellular and signaling pathway studies.