Learn More
Medchemexpress LLC PF-562271 besylate | 939791-38-5 | MFCD14105612 | 99.2% | 665.66 g/mol | C27H26F3N7O6S2 | 1 ML
SDP
Catalog No. 5000243406
Encompass_Preferred
PF-562271 besylate is a potent, ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 used in biochemical, cell-based, and in vivo research. It is supplied as a 10 mM solution in DMSO and exhibits low-nanomolar activity against FAK and nanomolar activity against Pyk2.
  • Potent inhibition of FAK and Pyk2 (FAK IC50 = 1.5 nM; Pyk2 IC50 = 13 nM).
  • Supplied as a ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High reported purity suitable for research use (99.17%).
  • Demonstrated antiproliferative activity across multiple cancer cell lines.
  • Validated pharmacological effects in vivo, including inhibition of FAK phosphorylation.
Catalog No. 50-002-43406 Supplier Medchemexpress LLC Supplier No. HY1045810MM 1ML
PF-562271 besylate is a potent, ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 used in biochemical, cell-based, and in vivo research. It is supplied as a 10 mM solution in DMSO and exhibits low-nanomolar activity against FAK and nanomolar activity against Pyk2.
  • Potent inhibition of FAK and Pyk2 (FAK IC50 = 1.5 nM; Pyk2 IC50 = 13 nM).
  • Supplied as a ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High reported purity suitable for research use (99.17%).
  • Demonstrated antiproliferative activity across multiple cancer cell lines.
  • Validated pharmacological effects in vivo, including inhibition of FAK phosphorylation.