Learn More
Medchemexpress LLC PF-562271 besylate | 939791-38-5 | MFCD14105612 | 99.2% | 665.66 g·mol⁻¹ | C27H26F3N7O6S2 | 25 MG
SDP
Catalog No. 5000243274
Encompass_Preferred
PF-562271 besylate is a small-molecule, ATP-competitive and reversible inhibitor of focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (Pyk2). It exhibits low-nanomolar potency in biochemical assays and is used in biochemical and cell-based studies to investigate FAK/Pyk2 signaling, cell migration, and cancer-related pathways. Supplied as a high-purity solid for research use.
  • Potent ATP-competitive, reversible inhibition of FAK and Pyk2.
  • Low-nanomolar biochemical potency (FAK IC50 ≈ 1.5 nM; Pyk2 IC50 ≈ 13 nM).
  • Suitable for biochemical and cell-based assays investigating signaling and migration.
  • High purity appropriate for research applications.
  • Soluble in DMSO and largely insoluble in water.
  • Available in small research pack sizes for laboratory studies.
Catalog No. 50-002-43274 Supplier Medchemexpress LLC Supplier No. HY1045825MG
PF-562271 besylate is a small-molecule, ATP-competitive and reversible inhibitor of focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (Pyk2). It exhibits low-nanomolar potency in biochemical assays and is used in biochemical and cell-based studies to investigate FAK/Pyk2 signaling, cell migration, and cancer-related pathways. Supplied as a high-purity solid for research use.
  • Potent ATP-competitive, reversible inhibition of FAK and Pyk2.
  • Low-nanomolar biochemical potency (FAK IC50 ≈ 1.5 nM; Pyk2 IC50 ≈ 13 nM).
  • Suitable for biochemical and cell-based assays investigating signaling and migration.
  • High purity appropriate for research applications.
  • Soluble in DMSO and largely insoluble in water.
  • Available in small research pack sizes for laboratory studies.