Learn More
Medchemexpress LLC PF-562271 besylate | 939791-38-5 | MFCD14105612 | 99.2% | 665.7 g/mol | C27H26F3N7O6S2 | 1 MG
SDP
Catalog No. 5000243213
Encompass_Preferred
PF-562271 besylate is an ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (Pyk2) used as a research tool in cell-signaling and oncology studies. It exhibits potent biochemical activity (FAK IC50 ≈ 1.5 nM; Pyk2 IC50 ≈ 13-14 nM), is supplied as the besylate salt, and is available at high purity for in vitro and in vivo research.
  • Potent ATP-competitive, reversible inhibitor of FAK and Pyk2.
  • Low nanomolar biochemical potency for target kinases.
  • Provided as a besylate salt for improved stability and handling.
  • High reported purity suitable for research applications.
  • Used in cellular and animal model studies of tumor signaling pathways.
Catalog No. 50-002-43213 Supplier Medchemexpress LLC Supplier No. HY104581MG
PF-562271 besylate is an ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (Pyk2) used as a research tool in cell-signaling and oncology studies. It exhibits potent biochemical activity (FAK IC50 ≈ 1.5 nM; Pyk2 IC50 ≈ 13-14 nM), is supplied as the besylate salt, and is available at high purity for in vitro and in vivo research.
  • Potent ATP-competitive, reversible inhibitor of FAK and Pyk2.
  • Low nanomolar biochemical potency for target kinases.
  • Provided as a besylate salt for improved stability and handling.
  • High reported purity suitable for research applications.
  • Used in cellular and animal model studies of tumor signaling pathways.