Learn More
Medchemexpress LLC PF-562271 besylate | 939791-38-5 | MFCD16038299 | 99.2% | 665.66 g·mol⁻¹ | C27H26F3N7O6S2 | 5 MG
SDP
Catalog No. 5000242580
Encompass_Preferred
PF-562271 besylate is a small-molecule, ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 used in research to study FAK pathway modulation. It is supplied as the besylate salt for laboratory use and has been evaluated in preclinical and clinical studies.
  • Potent ATP-competitive inhibitor of FAK and Pyk2 (IC50 1.5 nM and 13 nM).
  • Reversible mechanism suitable for biochemical and cellular assays.
  • Supplied as besylate salt for improved stability and handling.
  • High chemical purity (99.17%) for reproducible results.
  • Molecular weight 665.66 g·mol⁻¹, molecular formula C27H26F3N7O6S2.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-42580 Supplier Medchemexpress LLC Supplier No. HY104585MG
PF-562271 besylate is a small-molecule, ATP-competitive, reversible inhibitor of focal adhesion kinase (FAK) and Pyk2 used in research to study FAK pathway modulation. It is supplied as the besylate salt for laboratory use and has been evaluated in preclinical and clinical studies.
  • Potent ATP-competitive inhibitor of FAK and Pyk2 (IC50 1.5 nM and 13 nM).
  • Reversible mechanism suitable for biochemical and cellular assays.
  • Supplied as besylate salt for improved stability and handling.
  • High chemical purity (99.17%) for reproducible results.
  • Molecular weight 665.66 g·mol⁻¹, molecular formula C27H26F3N7O6S2.
  • Intended for research use only; not for human or veterinary use.