Learn More
Selleck Chemical LLC PF-562271 HCl
SDP
Supplier:  Selleck Chemical LLC S735725MG
Encompass_Preferred
PF-562271 HCl is the hydrochloride salt of PF-562271 which is a potent ATP-competitive reversible inhibitor of FAK with IC50 of 1 5 nM 10-fold less potent for Pyk2 than FAK and 100-fold selectivity against other protein kinases except for some CDKs Phase 1
Catalog No. 50-000-04360