Learn More
Apexbio Technology LLC PF-562271 HCl, 50mg. Cas: 939791-41-0 MFCD: MFCD17170115. FAK/Pyk2 inhibitor
SDP
Catalog No. 501905121
Encompass_Preferred
PF-562271 is a potent, ATP-competitive and reversible inhibitor of both focal adhesion kinase (FAK), a non-receptor tyrosine kinase involved in a variety of cellular events, and proline-rich tyrosine kinase 2 (Pyk2), an FAK homolog containing 48% amino acid identity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5121 Supplier Apexbio Technology LLC Supplier No. A834550
PF-562271 is a potent, ATP-competitive and reversible inhibitor of both focal adhesion kinase (FAK), a non-receptor tyrosine kinase involved in a variety of cellular events, and proline-rich tyrosine kinase 2 (Pyk2), an FAK homolog containing 48% amino acid identity. Other sizes are available. Please inqury us for quote.