Learn More
Cayman Chemical PF-562271 (besylate), Purity: ≥98%, CAS: 939791-38-5, 50 mg
SDP
Catalog No. 502965991
Encompass_Preferred
An ATP-competitive, reversible inhibitor of FAK (IC50 = 1.5 nM) and PYK2 (IC50 = 14 nM); inhibits FAK phosphorylation (EC50 = 93 ng/ml) in glioblastoma-bearing mice and has been shown to regress tumors in multiple xenograft models
Catalog No. 50-296-5991 Supplier Cayman Chemical Supplier No. 1849950 MG
An ATP-competitive, reversible inhibitor of FAK (IC50 = 1.5 nM) and PYK2 (IC50 = 14 nM); inhibits FAK phosphorylation (EC50 = 93 ng/ml) in glioblastoma-bearing mice and has been shown to regress tumors in multiple xenograft models