Learn More
Abcam PF-573228, 10MG
Catalog No. 502671876
Encompass_Preferred
MW 491.5 Da, Purity >=98%. Potent and selective FAK inhibitor (IC₅₀ = 4 nM versus recombunant catalytic fragent). Blocks FAK Tyr397phosphorylation in cultured cells (IC₅₀ = 30–100 nM). Shows 50 - 250-fold selectivity for FAK aginst a panel of 40 recombinant kinases. Inhibits chemotactic and haptotactic cell migration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1876 Supplier Abcam Supplier No. AB27352310MG
MW 491.5 Da, Purity >=98%. Potent and selective FAK inhibitor (IC₅₀ = 4 nM versus recombunant catalytic fragent). Blocks FAK Tyr397phosphorylation in cultured cells (IC₅₀ = 30–100 nM). Shows 50 - 250-fold selectivity for FAK aginst a panel of 40 recombinant kinases. Inhibits chemotactic and haptotactic cell migration.
The product is subject to the following: Abcam Restricted Use Statement