Learn More
TARGETMOL CHEMICALS INC PF-573228 25MG
SDP
Catalog No. 502577250
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-573228 is an ATP-competitive FAK inhibitor. In a cell-free assay the IC50 of FAK is 4 nM. purity: 98%
Catalog No. 50-257-7250 Supplier TARGETMOL CHEMICALS INC Supplier No. T200125MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-573228 is an ATP-competitive FAK inhibitor. In a cell-free assay the IC50 of FAK is 4 nM. purity: 98%