Learn More
Medchemexpress LLC HY-10461 50mg Medchemexpress, PF-573228 CAS:869288-64-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1046150MG
Encompass_Preferred
Medchemexpress, HY-10461 50mg PF-573228 CAS:869288-64-2 PF-573228 is a potent and selective FAK inhibitor with IC50 of 4 nM for purified recombinant catalytic fragment of FAK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8227