Learn More
Enzo Life Sciences PF-622 (5mg). CAS: 898235-65-9
SDP
Catalog No. 502010781
Encompass_Preferred
Novel, potent and selective inhibitor of fatty acid amide hydrolase (FAAH). Inhibits FAAH in a time-dependent manner by covalently modifying the enzyme's active site serine (IC50=33nM). It is completely selective for FAAH relative to other mammalian serine hydrolases. Alternative name: N-phenyl-4-(quinolin-2-ylmethyl)piperazine-1-carboxamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (7mg/ml) or 100% ethanol (3mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0781 Supplier Enzo Life Sciences Supplier No. BMLEI4020005
Novel, potent and selective inhibitor of fatty acid amide hydrolase (FAAH). Inhibits FAAH in a time-dependent manner by covalently modifying the enzyme's active site serine (IC50=33nM). It is completely selective for FAAH relative to other mammalian serine hydrolases. Alternative name: N-phenyl-4-(quinolin-2-ylmethyl)piperazine-1-carboxamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (7mg/ml) or 100% ethanol (3mg/ml). Long Term Storage: Ambient.