Learn More
Apexbio Technology LLC PF-670462 950912-80-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063242
Encompass_Preferred
PF-670462 (CAS 950912-80-8) is a selective inhibitor of casein kinase 1 (CK1) isoforms CK1 and CK1 serine/threonine kinases involved in circadian rhythm regulation It exhibits inhibitory activity toward CK1 and CK1 with IC50 values of approximately 80 nM and 13 nM respectively In cellular models co-transfected with GFP-tagged PER3 and human CK1 PF-670462 blocks nuclear translocation of PER3 In primary fibroblasts from wild-type mice it dose-dependently prolongs circadian cycle length PF-670462 serves as a useful chemical tool for studying CK1-mediated modulation of circadian rhythms
Catalog No. 50-000-63242 Supplier Apexbio Technology LLC Supplier No. A37195.1
PF-670462 (CAS 950912-80-8) is a selective inhibitor of casein kinase 1 (CK1) isoforms CK1 and CK1 serine/threonine kinases involved in circadian rhythm regulation It exhibits inhibitory activity toward CK1 and CK1 with IC50 values of approximately 80 nM and 13 nM respectively In cellular models co-transfected with GFP-tagged PER3 and human CK1 PF-670462 blocks nuclear translocation of PER3 In primary fibroblasts from wild-type mice it dose-dependently prolongs circadian cycle length PF-670462 serves as a useful chemical tool for studying CK1-mediated modulation of circadian rhythms