Learn More
Abcam PF 670462, casein kinase 1epsilon (CK1epsilon) and CK1delta inhibitor, 50MG
Catalog No. 502707059
Encompass_Preferred
MW 410.3 Da, Purity >99%. Potent, selective casein kinase 1ε (CK1ε) and CK1δ inhibitor (IC₅₀ values are 7.7 and 14 nM for CK1ε and CK1δ, respectively). Shows >300-fold selectivity over CK2 and selectivity over more than 40 other kinases. Alters circadian rhythms in vivo. Also available in simple stock solutions (ab146728) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7059 Supplier Abcam Supplier No. AB12098550MG
MW 410.3 Da, Purity >99%. Potent, selective casein kinase 1ε (CK1ε) and CK1δ inhibitor (IC₅₀ values are 7.7 and 14 nM for CK1ε and CK1δ, respectively). Shows >300-fold selectivity over CK2 and selectivity over more than 40 other kinases. Alters circadian rhythms in vivo. Also available in simple stock solutions (ab146728) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement