Learn More
Medchemexpress LLC PF-670462 dihydrochloride | 950912-80-8 | C19H22Cl2FN5 | 5 MG
SDP
Catalog No. 5000376084
Encompass_Preferred
PF-670462 dihydrochloride is a potent and selective inhibitor of casein kinase (CK1ε and CK1δ) and a potent inhibitor of Wnt/β-catenin signaling. In vivo, it produces robust phase delays in circadian rhythms, affecting gene transcription in the liver, pancreas, and suprachiasmatic nucleus.
  • Potent and selective inhibitor of CKIε and CKIδ
  • Potent inhibitor of Wnt/β-catenin signaling
  • Lower selectivity for EGFR and SAPK2A/p38
  • Causes GFP signal redistribution to the cytoplasm
  • Produces robust phase delays in circadian rhythms in rats
  • Delays rhythmic transcription of Bmal1, Per1, Per2, and Nr1d1 in liver and pancreas
  • Delays Nr1d1 and Dbp mRNA expression in the suprachiasmatic nucleus
Catalog No. 50-003-76084 Supplier Medchemexpress LLC Supplier No. HY154905MG
PF-670462 dihydrochloride is a potent and selective inhibitor of casein kinase (CK1ε and CK1δ) and a potent inhibitor of Wnt/β-catenin signaling. In vivo, it produces robust phase delays in circadian rhythms, affecting gene transcription in the liver, pancreas, and suprachiasmatic nucleus.
  • Potent and selective inhibitor of CKIε and CKIδ
  • Potent inhibitor of Wnt/β-catenin signaling
  • Lower selectivity for EGFR and SAPK2A/p38
  • Causes GFP signal redistribution to the cytoplasm
  • Produces robust phase delays in circadian rhythms in rats
  • Delays rhythmic transcription of Bmal1, Per1, Per2, and Nr1d1 in liver and pancreas
  • Delays Nr1d1 and Dbp mRNA expression in the suprachiasmatic nucleus