Learn More
Cayman Chemical PF-670462 (hydrochloride), Purity: ≥99%, CAS: 950912-80-8, 50 mg
SDP
Catalog No. 502969390
Encompass_Preferred
A potent inhibitor of the CK1 isoforms CK1ε and CK1δ (IC50 = 7.7 and 14 nM, respectively); less effectively inhibits a wide variety of related or common kinases; disrupts circadian rhythms in cells and animals; blocks the locomotor response to amphetamines in mice
Catalog No. 50-296-9390 Supplier Cayman Chemical Supplier No. 1458850 MG
A potent inhibitor of the CK1 isoforms CK1ε and CK1δ (IC50 = 7.7 and 14 nM, respectively); less effectively inhibits a wide variety of related or common kinases; disrupts circadian rhythms in cells and animals; blocks the locomotor response to amphetamines in mice