Learn More
Medchemexpress LLC Pf-794 | 2056111-45-4 | 99.2% | 100 MG
SDP
Catalog No. 5000230177
Encompass_Preferred
Pf-794 is a small-molecule research compound that functions as a selective, ATP-competitive inhibitor of Traf2- and Nck-interacting kinase (TNIK). It is supplied for laboratory research and preclinical studies.
  • Targets Traf2- and Nck-interacting kinase (TNIK).
  • ATP-competitive mechanism of inhibition.
  • Reported high purity for reliable experimental use.
  • Available in small research pack sizes commonly 100 mg.
  • CAS number 2056111-45-4 for unambiguous identification.
Catalog No. 50-002-30177 Supplier Medchemexpress LLC Supplier No. HY122805100MG
Pf-794 is a small-molecule research compound that functions as a selective, ATP-competitive inhibitor of Traf2- and Nck-interacting kinase (TNIK). It is supplied for laboratory research and preclinical studies.
  • Targets Traf2- and Nck-interacting kinase (TNIK).
  • ATP-competitive mechanism of inhibition.
  • Reported high purity for reliable experimental use.
  • Available in small research pack sizes commonly 100 mg.
  • CAS number 2056111-45-4 for unambiguous identification.