Learn More
Medchemexpress LLC Pf-794 | 2056111-45-4 | 99.2% | 25 MG
SDP
Catalog No. 5000230425
Encompass_Preferred
PF-794 is a potent, selective, ATP-competitive Traf2- and Nck-interacting kinase (TNIK) inhibitor (IC50 39 nM) used as a research tool in neuroscience and signal transduction studies. It reduces endogenous p120-catenin phosphorylation in cells and is supplied as a high-purity research chemical.
  • Potent TNIK inhibitor with IC50 of 39 nM.
  • Selective for the TNIK family.
  • Reduces endogenous p120-catenin phosphorylation in cells.
  • High reported purity (99.19%).
  • Molecular formula C22H20N4O; molecular weight 356.42 g/mol.
Catalog No. 50-002-30425 Supplier Medchemexpress LLC Supplier No. HY12280525MG
PF-794 is a potent, selective, ATP-competitive Traf2- and Nck-interacting kinase (TNIK) inhibitor (IC50 39 nM) used as a research tool in neuroscience and signal transduction studies. It reduces endogenous p120-catenin phosphorylation in cells and is supplied as a high-purity research chemical.
  • Potent TNIK inhibitor with IC50 of 39 nM.
  • Selective for the TNIK family.
  • Reduces endogenous p120-catenin phosphorylation in cells.
  • High reported purity (99.19%).
  • Molecular formula C22H20N4O; molecular weight 356.42 g/mol.