Learn More
Apexbio Technology LLC PF-8380, 10mg. Cas: 1144035-53-9 MFCD: MFCD20527274. Autotaxin inhibitor,potent and specific
SDP
Supplier:  Apexbio Technology LLC A372010
Encompass_Preferred
PF-8380 is a specific and potent inhibitor of Autotaxin (ATX) with an IC50 value of 2.8 nM [1].Autotaxin (ATX) is a secreted enzyme having lysophospholipase D activity Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4831