Learn More
Selleck Chemical LLC PF-8380 200mg 1144035-53-9
SDP
Supplier:  Selleck Chemical LLC S8218200MG
Encompass_Preferred
PF-8380 is a potent autotaxin (ATX) inhibitor with IC50 of 2.8 nM in an in vitro enzyme assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5812