Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-8380 hydrochloride | 2070015-01-7 | 98.6% | 514.79 g/mol | C22H22ClN2O5S | 5 MG
SDP

Catalog No. 5000262684
Encompass_Preferred

PF-8380 hydrochloride is a small-molecule autotaxin (ATX) inhibitor supplied as a research reagent. It demonstrates potent inhibition in isolated enzyme assays and measurable activity in human whole blood, and is intended for laboratory research use only.

  • Potent autotaxin inhibitor; IC50 2.8 nM in isolated enzyme assay.
  • Active in human whole blood with IC50 ~101 nM.
  • Molecular weight 514.79 g/mol and formula C22H22ClN2O5S.
  • High purity (98.6%), suitable for biochemical and cellular assays.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-62684 Supplier Medchemexpress LLC Supplier No. HY13344A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-8380 hydrochloride is a small-molecule autotaxin (ATX) inhibitor supplied as a research reagent. It demonstrates potent inhibition in isolated enzyme assays and measurable activity in human whole blood, and is intended for laboratory research use only.

  • Potent autotaxin inhibitor; IC50 2.8 nM in isolated enzyme assay.
  • Active in human whole blood with IC50 ~101 nM.
  • Molecular weight 514.79 g/mol and formula C22H22ClN2O5S.
  • High purity (98.6%), suitable for biochemical and cellular assays.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.