Learn More
Medchemexpress LLC HY-132283 5mg , PF-9363 CAS:2569009-58-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1322835MG
Encompass_Preferred
HY-132283 5mg PF-9363 CAS: 2569009-58-9 PF-9363 (CTx-648) is a first-in-class potent and high selective KAT6A/KAT6B inhibitor. PF-9363 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9689