Learn More
Abcam PF-9366, 5MG
Supplier:  Abcam AB2869645MG
Encompass_Preferred
MW 350.8 Da, Purity >=98%. An allosteric inhibitor of human methionine adenosyltransferase 2A (Mat2A), the extrahepatic isoform (IC₅₀ = 420 nM). PF-9366 inhibits cellular S-Adenosyl-L-methionine (SAM) production with an IC₅₀ of 1.2 μM in H520 lung carcinoma cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5807