Learn More
Medchemexpress LLC HY-10721 5mg Medchemexpress, PF-AKT400 CAS:1004990-28-6 Purity:>98%
SDP
Catalog No. 501968370
Encompass_Preferred
Medchemexpress, HY-10721 5mg PF-AKT400 CAS:1004990-28-6 PF-AKT400 is a broadly selective, potent, ATP-competitive Akt inhibitor, displays 900-fold greater selectivity for PKBα (IC50=0.5 nM) than PKA (IC50=450 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8370 Supplier Medchemexpress LLC Supplier No. HY107215MG
Medchemexpress, HY-10721 5mg PF-AKT400 CAS:1004990-28-6 PF-AKT400 is a broadly selective, potent, ATP-competitive Akt inhibitor, displays 900-fold greater selectivity for PKBα (IC50=0.5 nM) than PKA (IC50=450 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.