Learn More
TARGETMOL CHEMICALS INC PF07104091 5MG
SDP
Catalog No. 502578970
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF07104091 inhibits CDK2 which may lead to cell cycle arrest induce apoptosis and inhibit tumor cell proliferation. purity: 98%
Catalog No. 50-257-8970 Supplier TARGETMOL CHEMICALS INC Supplier No. T97125MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF07104091 inhibits CDK2 which may lead to cell cycle arrest induce apoptosis and inhibit tumor cell proliferation. purity: 98%