Learn More
Accuflow Systems Saint Gobain Retainer, PVDF, 1/4 in. I.D., 250/Pk.
Catalog No. NC3440519
Encompass_Preferred
This retainer is a component of the Pure-Fit® SIB® family, designed to provide seamless transitions in fluid paths by eliminating entrapment and leak points. Made from PVDF, it ensures robust and biocompatible fluid connections.
  • Smooth inner bore design
  • Eliminates entrapment and leak points
  • Made of PVDF material
  • Meets USP <88> Class VI, USP <87>, and/or ISO 10993-5 for biocompatibility
  • Internal diameter of 1/4 in. (6.4 mm)
Catalog No. NC3440519 Supplier Accuflow Systems Supplier No. PF135250PVDF
This retainer is a component of the Pure-Fit® SIB® family, designed to provide seamless transitions in fluid paths by eliminating entrapment and leak points. Made from PVDF, it ensures robust and biocompatible fluid connections.
  • Smooth inner bore design
  • Eliminates entrapment and leak points
  • Made of PVDF material
  • Meets USP <88> Class VI, USP <87>, and/or ISO 10993-5 for biocompatibility
  • Internal diameter of 1/4 in. (6.4 mm)