Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Retainer, PVDF, 3/8 in
Catalog No. NC3440522
Encompass_Preferred
Saint-Gobain Pure-Fit SIB retainer manufactured from PVDF for smooth inner bore (SIB) fluid connections. The retainer provides 360° radial compression behind the barb to create a leak-free transition between tubing and fittings, minimizing dead volume and potential sites for contamination. Designed for bioprocess and laboratory fluid transfer, the component is cleanroom packaged and commonly supplied in bags of 250.
  • Smooth inner bore design eliminates dead volume and reduces entrapment.
  • PVDF construction provides chemical resistance and temperature tolerance.
  • 3/8 in (9.5 mm) inner diameter, 360° radial compression for leak-free connections.
  • Suitable for bioprocessing and sterile fluid transfer; meets USP Class VI and relevant ISO criteria.
  • Manufactured and packaged in a certified cleanroom; typically packaged 250 per bag.
Catalog No. NC3440522 Supplier Accuflow Systems Supplier No. PF135375PVDF
Saint-Gobain Pure-Fit SIB retainer manufactured from PVDF for smooth inner bore (SIB) fluid connections. The retainer provides 360° radial compression behind the barb to create a leak-free transition between tubing and fittings, minimizing dead volume and potential sites for contamination. Designed for bioprocess and laboratory fluid transfer, the component is cleanroom packaged and commonly supplied in bags of 250.
  • Smooth inner bore design eliminates dead volume and reduces entrapment.
  • PVDF construction provides chemical resistance and temperature tolerance.
  • 3/8 in (9.5 mm) inner diameter, 360° radial compression for leak-free connections.
  • Suitable for bioprocessing and sterile fluid transfer; meets USP Class VI and relevant ISO criteria.
  • Manufactured and packaged in a certified cleanroom; typically packaged 250 per bag.