Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Retainer, PVDF, 1/2 in
Catalog No. NC3440524
Encompass_Preferred
Saint-Gobain Pure-Fit SIB retainer in PVDF provides a smooth inner-bore connection for 1/2 in (12.7 mm) flexible tubing. It is designed to minimize dead volume and deliver secure, leak-resistant hose-barb connections for bioprocessing and laboratory fluid transfer applications.
  • Retainer for 1/2 in (12.7 mm) tubing connections.
  • PVDF construction; autoclavable and gamma stable.
  • Designed to minimize dead volume and internal turbulence.
  • Provides 360° compression for leak-resistant connections.
  • Manufactured and packaged in a certified clean room.
Catalog No. NC3440524 Supplier Accuflow Systems Supplier No. PF135500PVDF
Saint-Gobain Pure-Fit SIB retainer in PVDF provides a smooth inner-bore connection for 1/2 in (12.7 mm) flexible tubing. It is designed to minimize dead volume and deliver secure, leak-resistant hose-barb connections for bioprocessing and laboratory fluid transfer applications.
  • Retainer for 1/2 in (12.7 mm) tubing connections.
  • PVDF construction; autoclavable and gamma stable.
  • Designed to minimize dead volume and internal turbulence.
  • Provides 360° compression for leak-resistant connections.
  • Manufactured and packaged in a certified clean room.