Learn More
Accuflow Systems Saint Gobain Retainer, PVDF, 3/4 in., 25/Pk.
Catalog No. NC3440527
Encompass_Preferred
Pure-Fit® SIB® retainers are designed to ensure a seamless and leak-free fluid path between tubing and fittings. Their unique internal design eliminates voids that can lead to turbulence or stagnation, preventing potential bacterial growth.
  • Seamless transition between tubing and fittings
  • Eliminates entrapment and leak points
  • Unique internal design eliminates voids to prevent bacterial growth
  • Provides complete 360° compression for leak-free connections
  • Meets USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements
  • Manufactured and packaged in a certified clean room
  • Made from animal-free, BPA-free PVDF material, which is autoclaveable and gamma stable
Catalog No. NC3440527 Supplier Accuflow Systems Supplier No. PF135750PVDFAL
Pure-Fit® SIB® retainers are designed to ensure a seamless and leak-free fluid path between tubing and fittings. Their unique internal design eliminates voids that can lead to turbulence or stagnation, preventing potential bacterial growth.
  • Seamless transition between tubing and fittings
  • Eliminates entrapment and leak points
  • Unique internal design eliminates voids to prevent bacterial growth
  • Provides complete 360° compression for leak-free connections
  • Meets USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements
  • Manufactured and packaged in a certified clean room
  • Made from animal-free, BPA-free PVDF material, which is autoclaveable and gamma stable