Learn More
Medchemexpress LLC Pf15 TFA | 99.7% | 969.97 | C46H50F3N13O8 | 5 MG
SDP
Catalog No. 5000220949
Encompass_Preferred
A heterobifunctional PROTAC that recruits cereblon to promote degradation of FLT3-ITD. It reduces FLT3 and STAT5 phosphorylation, inhibits proliferation of FLT3-ITD-positive cells, and shows tumor growth inhibition in preclinical models. Supplied as a trifluoroacetic acid salt for laboratory research.
Catalog No. 50-002-20949 Supplier Medchemexpress LLC Supplier No. HY143286A5MG
A heterobifunctional PROTAC that recruits cereblon to promote degradation of FLT3-ITD. It reduces FLT3 and STAT5 phosphorylation, inhibits proliferation of FLT3-ITD-positive cells, and shows tumor growth inhibition in preclinical models. Supplied as a trifluoroacetic acid salt for laboratory research.