Learn More
Medchemexpress LLC HY-13447A 5mg Medchemexpress, PF429242 (dihydrochloride) CAS:2248666-66-0 Purity:>98%
SDP
Catalog No. 502081368
Encompass_Preferred
Medchemexpress, HY-13447A 5mg PF429242 (dihydrochloride) CAS:2248666-66-0 PF429242 dihydrochloride is a reversible and competitive SREBP site 1 protease (S1P) inhibitor with an IC50 of 175 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1368 Supplier Medchemexpress LLC Supplier No. HY13447A5MG
Medchemexpress, HY-13447A 5mg PF429242 (dihydrochloride) CAS:2248666-66-0 PF429242 dihydrochloride is a reversible and competitive SREBP site 1 protease (S1P) inhibitor with an IC50 of 175 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.