Learn More
TARGETMOL CHEMICALS INC PF429242 dihydrochloride 10MG
SDP
Catalog No. 502879643
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. PF429242 dihydrochloride is a reversible and competitive inhibitor of S1P, with an IC50 of 175 nM. Purity 98.67%
Catalog No. 50-287-9643 Supplier TARGETMOL CHEMICALS INC Supplier No. T1243710MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. PF429242 dihydrochloride is a reversible and competitive inhibitor of S1P, with an IC50 of 175 nM. Purity 98.67%