Learn More
Aviva Systems Biology Corp PF4V1 Antibody - middle region (ARP42053_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42053_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PF4V1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PF4V1. Alternate names: CXCL4V1, PF4-ALT, PF4A, SCYB4V1, CXCL4L1ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-58093