Learn More
MSC Baldwin Fuel Filter Element, 15 micron, 5 1/16 in OD
Catalog No. 5000508301
Encompass
This fuel filter element is designed for modern fuel injection systems, which require fuel to be free of both particulate and water contamination. It helps keep fuel clean and engines running at maximum efficiency.
  • Keeps fuel clean.
  • Ensures engines run at maximum efficiency.
  • Features a cartridge filter design.
  • Provides a 15 micron absolute filter rating.
  • Outside diameter measures 5 1/16 in (128.6 mm).
  • Inside diameter measures 11/16 in (17.5 mm).
  • Overall length is 20 15/16 in (531.8 mm).
  • Includes an attached gasket and spring.
  • Suitable for railroad applications.
Catalog No. 50-005-08301 Supplier MSC Supplier No. 55866156
This fuel filter element is designed for modern fuel injection systems, which require fuel to be free of both particulate and water contamination. It helps keep fuel clean and engines running at maximum efficiency.
  • Keeps fuel clean.
  • Ensures engines run at maximum efficiency.
  • Features a cartridge filter design.
  • Provides a 15 micron absolute filter rating.
  • Outside diameter measures 5 1/16 in (128.6 mm).
  • Inside diameter measures 11/16 in (17.5 mm).
  • Overall length is 20 15/16 in (531.8 mm).
  • Includes an attached gasket and spring.
  • Suitable for railroad applications.